3-Chloro-2-methyl-benzoic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-chloro-2-methylbenzoate

A252364-1G A252364-250MG A252364-5G
77165-992EA 46.35 USD
77165-992 77165-994 77165-996
3-Chloro-2-methyl-benzoic acid methyl ester 98%
3-Chloro-2-methyl-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
MDL Number: MFCD00230148
CAS Number: 99586-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR