2,4-Dibromo-1,5-difluoro-3-nitrobenzene 95%

Supplier: Ambeed
Warning

A251142-250MG A251142-1G
77165-968EA 11 USD
77165-968 77892-462
2,4-Dibromo-1,5-difluoro-3-nitrobenzene 95%
2,4-Dibromo-1,5-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆HBr₂F₂NO₂
MDL Number: MFCD22681519
CAS Number: 144450-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR