Propyl Levulinate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Levulinic acid Propyl ester, 4-Oxovaleric acid Propyl ester, Propyl 4-Oxovalerate, 4-Oxopentanoic acid Propyl ester, Propyl 4-Oxopentanoate

A251008-10G A251008-25G A251008-5G A251008-1G
77165-962EA 36.05 USD
77165-962 77165-964 77165-966 77683-751
Propyl Levulinate 97%, Ambeed.Inc
Propyl Levulinate

Precaución: For research use only.

Formula: C₈H₁₄O₃
MW: 158.20 g/mol
MDL Number: MFCD16041935
CAS Number: 645-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR