3-Aminobenzhydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Aminobenzoic hydrazide, 3-Aminobenzene-1-carbohydrazide, 3-Aminobenzohydrazide, 3-Aminobenzoic acid hydrazide

A250602-100G A250602-25G A250602-5G
77193-636EA 152 USD
77193-636 77193-638 77193-640
3-Aminobenzhydrazide 98%
3-Aminobenzhydrazide

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MDL Number: MFCD00017067
CAS Number: 14062-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR