trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (trans-4-(2-hydroxyethyl)cyclohexyl)carbamate
A250414-1G
A250414-250MG
A250414-5G
A250414-100MG
77165-950EA
104.03
USD
77165-950
77165-952
77165-954
77683-749
trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane 97%
trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane
Precaución: For research use only.
Formula:
C₁₃H₂₅NO₃ MW: 243.347 g/mol |
MDL Number:
MFCD03844626 CAS Number: 917342-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...