5-Bromo-2-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A250305-100G A250305-1G A250305-5G A250305-25G
77193-630EA 274.99 USD
77193-630 77193-632 77193-634 77304-518
5-Bromo-2-methoxyphenylboronic acid 97%
5-Bromo-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
MDL Number: MFCD00093409
CAS Number: 89694-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR