1,5-Dichloro-3-methoxy-2-nitrobenzene 95%

Supplier: Ambeed
Warning

A249922-1G A249922-250MG A249922-5G
77165-942EA 293 USD
77165-942 77165-944 77165-946
1,5-Dichloro-3-methoxy-2-nitrobenzene 95%
1,5-Dichloro-3-methoxy-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₃
MDL Number: MFCD28134572
CAS Number: 74672-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR