2-((6-Amino-1H-purin-1-yl)methoxy)ethanol 97%

Supplier: Ambeed
Warning

A249379-100MG A249379-250MG
77165-938EA 58 USD
77165-938 77165-940
2-((6-Amino-1H-purin-1-yl)methoxy)ethanol 97%
2-((6-Amino-1H-purin-1-yl)methoxy)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₁N₅O₂
MDL Number: MFCD11505837
CAS Number: 31383-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR