(Z)-Methyl 2-acetamido-3-phenylacrylate 95%

Supplier: Ambeed
Warning

A248557-1G A248557-250MG A248557-5G
77193-602EA 105.06 USD
77193-602 77193-604 77193-606
(Z)-Methyl 2-acetamido-3-phenylacrylate 95%
(Z)-Methyl 2-acetamido-3-phenylacrylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.23 g/mol
MDL Number: MFCD00452767
CAS Number: 60676-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR