JH-II-127 98+%

Supplier: Ambeed
Warning

Synonyms: (4-((5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl)amino)-3-methoxyphenyl)(morpholino)methanone

A248253-10MG A248253-1MG A248253-2MG A248253-5MG
77193-594EA 176.13 USD
77193-594 77193-596 77193-598 77193-600
JH-II-127 98+%
JH-II-127

Precaución: for Research Use Only

Formula: C₁₉H₂₁ClN₆O₃
MW: 416.87 g/mol
MDL Number: MFCD28975160
CAS Number: 1700693-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR