1,2,4,5-Tetrabromobenzene 98%

Supplier: Ambeed
Warning

A248034-100G A248034-1G A248034-25G A248034-5G
77193-578EA 183.34 USD
77193-578 77193-580 77193-582 77193-584
1,2,4,5-Tetrabromobenzene 98%
1,2,4,5-Tetrabromobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₄
MW: 393.7 g/mol
MDL Number: MFCD00000063
CAS Number: 636-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR