(S)-(+)-1-Phenyl-1,2-ethanediol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-Phenyl-1,2-ethanediol, (S)-1-Phenylethane-1,2-diol, (S)-(+)-Phenylethane-1,2-diol, (S)-(+)-α,β-Dihydroxyethylbenzene

A247722-100G A247722-10G A247722-1G A247722-25G
77193-556EA 298 USD
77193-556 77193-558 77193-560 77193-562
(S)-(+)-1-Phenyl-1,2-ethanediol 98%
(S)-(+)-1-Phenyl-1,2-ethanediol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MDL Number: MFCD00066256
CAS Number: 25779-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR