1-dodecyl-3-methylimidazolium tetrafluoroborate 99%

Supplier: Ambeed
Warning

A247556-1G A247556-25G A247556-5G
77193-546EA 6.18 USD
77193-546 77193-548 77193-550
1-dodecyl-3-methylimidazolium tetrafluoroborate 99%
1-dodecyl-3-methylimidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₆H₃₁BF₄N₂
MW: 338.24 g/mol
MDL Number: MFCD12022374
CAS Number: 244193-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR