3-Hydroxyquinoline-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A247110-100MG A247110-1G A247110-250MG A247110-5G
77193-518EA 167 USD
77193-518 77193-520 77193-522 77193-524
3-Hydroxyquinoline-2-carboxylic acid 97%
3-Hydroxyquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MDL Number: MFCD08705721
CAS Number: 15462-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR