6-Amino-2-cyclopropylpyrimidin-4-ol 97%
Supplier: Ambeed
|
Warning
|
A246617-100MG
A246617-250MG
77165-902EA
17.51
USD
77165-902
77165-904
6-Amino-2-cyclopropylpyrimidin-4-ol 97%
6-Amino-2-cyclopropylpyrimidin-4-ol
Precaución: for Research Use Only
Formula:
C₇H₉N₃O |
MDL Number:
MFCD16988316 CAS Number: 954388-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...