Methyl 4-(3-methoxyphenyl)-2,4-dioxobutanoate 97%

Supplier: Ambeed
Warning

A246588-1G A246588-250MG
77193-514EA 121.54 USD
77193-514 77193-516
Methyl 4-(3-methoxyphenyl)-2,4-dioxobutanoate 97%
Methyl 4-(3-methoxyphenyl)-2,4-dioxobutanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₅
MW: 236.22 g/mol
MDL Number: MFCD04967394
CAS Number: 501653-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR