tert-Butyl 1H-pyrazol-3-ylcarbamate 95%
Supplier: Ambeed
|
Warning
|
A246110-100MG
A246110-1G
A246110-250MG
A246110-5G
77165-892EA
52.53
USD
77165-892
77165-894
77165-896
77683-738
tert-Butyl 1H-pyrazol-3-ylcarbamate 95%
tert-Butyl 1H-pyrazol-3-ylcarbamate
Precaución: For research use only.
Formula:
C₈H₁₃N₃O₂ MW: 183.20 g/mol |
MDL Number:
MFCD14635783 CAS Number: 952674-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...