2-Ethylbenzene-1,4-diol 95%

Supplier: Ambeed
Warning

A245785-100MG A245785-250MG A245785-1G
77193-510EA 21.63 USD
77193-510 77193-512 77746-380
2-Ethylbenzene-1,4-diol 95%
2-Ethylbenzene-1,4-diol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.16 g/mol
MDL Number: MFCD00017398
CAS Number: 2349-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR