(S)-2-Amino-3-methoxypropanoic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A245547-1G
A245547-25G
A245547-5G
77193-504EA
13
USD
77193-504
77193-506
77193-508
(S)-2-Amino-3-methoxypropanoic acid hydrochloride 95%
(S)-2-Amino-3-methoxypropanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₀ClNO₃ |
MDL Number:
MFCD03840366 CAS Number: 336100-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...