1-((1H-Pyrazol-4-yl)methyl)-4-methyl-5-((4-((6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
MI-503
A245515-10MG
A245515-1MG
A245515-25MG
A245515-2MG
A245515-5MG
77165-872EA
75.19
USD
77165-872
77165-874
77165-876
77165-878
77165-880
1-((1H-Pyrazol-4-yl)methyl)-4-methyl-5-((4-((6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile 99+%
1-((1H-Pyrazol-4-yl)methyl)-4-methyl-5-((4-((6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile
Precaución: for Research Use Only
Formula:
C₂₈H₂₇F₃N₈S MW: 564.63 g/mol |
MDL Number:
MFCD28506303 CAS Number: 1857417-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...