1,5-Pentanediol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pentane-1,5-diol, 1,5-Dihydroxy pentane
A245468-100G
A245468-25G
A245468-500G
77193-490EA
7.21
USD
77193-490
77193-492
77193-494
1,5-Pentanediol 95%
1,5-Pentanediol
Precaución: for Research Use Only
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol |
MDL Number:
MFCD00002978 CAS Number: 111-29-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...