4-Aminopyridin-2-ol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-pyridin-2-ol, 4-Amino-1H-pyridin-2-one

A245296-1G A245296-250MG
77165-856EA 37 USD
77165-856 77165-858
4-Aminopyridin-2-ol 97%
4-Aminopyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MDL Number: MFCD02682913
CAS Number: 59315-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR