5-Bromo-2-nitrothiazole 97%

Supplier: Ambeed
Warning

A245184-100MG A245184-250MG A245184-5G A245184-1G
77193-474EA 66 USD
77193-474 77193-476 77193-478 77593-560
5-Bromo-2-nitrothiazole 97%
5-Bromo-2-nitrothiazole

Precaución: for Research Use Only

Formula: C₃HBrN₂O₂S
MDL Number: MFCD28142694
CAS Number: 182692-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR