4-(2,4-Dichloro-5-pyrimidyl)morpholine 95%

Supplier: Ambeed
Warning

A245128-1G A245128-250MG
77165-850EA 157 USD
77165-850 77165-852
4-(2,4-Dichloro-5-pyrimidyl)morpholine 95%
4-(2,4-Dichloro-5-pyrimidyl)morpholine

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N₃O
MDL Number: MFCD20483716
CAS Number: 280582-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR