4-Methylpiperidin-2-one 98%

Supplier: Ambeed
Warning

A244570-100MG A244570-1G A244570-250MG A244570-5G
77165-842EA 52 USD
77165-842 77165-844 77165-846 77892-441
4-Methylpiperidin-2-one 98%
4-Methylpiperidin-2-one

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MDL Number: MFCD13178698
CAS Number: 4720-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR