5-((1,4-Diazepan-1-yl)sulfonyl)isoquinolin-1(2H)-one 98%

Supplier: Ambeed
Warning

A244270-10MG A244270-25MG A244270-50MG A244270-5MG A244270-1MG
77193-454EA 78.01 USD
77193-454 77193-456 77193-458 77193-460 77616-006
5-((1,4-Diazepan-1-yl)sulfonyl)isoquinolin-1(2H)-one 98%
5-((1,4-Diazepan-1-yl)sulfonyl)isoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₄H₁₇N₃O₃S
MDL Number: MFCD01752417
CAS Number: 105628-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR