1-((2R,4S,5R)-4-Hydroxy-5-((trityloxy)methyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 98% (contains < 10% DCM)

Supplier: Ambeed
Warning

A243879-10G A243879-1G A243879-250MG A243879-25G
77193-442EA 82.4 USD
77193-442 77193-444 77193-446 77746-371
1-((2R,4S,5R)-4-Hydroxy-5-((trityloxy)methyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 98% (contains < 10% DCM)
1-((2R,4S,5R)-4-Hydroxy-5-((trityloxy)methyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₂₉H₂₈N₂O₅
MW: 484.54 g/mol
MDL Number: MFCD00038462
CAS Number: 7791-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR