3-Bromo-5-nitrobenzenesulfonamide 97%
Supplier: Ambeed
|
Warning
|
A243872-100MG
A243872-1G
A243872-250MG
77193-436EA
56.65
USD
77193-436
77193-438
77193-440
3-Bromo-5-nitrobenzenesulfonamide 97%
3-Bromo-5-nitrobenzenesulfonamide
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₄S MW: 281.08 g/mol |
MDL Number:
MFCD18392269 CAS Number: 59481-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...