Benzyl 2-(((benzyloxy)carbonyl)amino)acrylate 95%

Supplier: Ambeed
Warning

A243794-1G A243794-250MG
77165-822EA 143.17 USD
77165-822 77165-824
Benzyl 2-(((benzyloxy)carbonyl)amino)acrylate 95%
Benzyl 2-(((benzyloxy)carbonyl)amino)acrylate

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄
MW: 311.33 g/mol
MDL Number: MFCD28369506
CAS Number: 59524-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR