1,2-Dimethylpiperazine 97%

Supplier: Ambeed
Danger

A243128-100MG A243128-10G A243128-1G A243128-250MG A243128-5G
77193-420EA 14.42 USD
77193-420 77193-422 77193-424 77193-426 77193-428
1,2-Dimethylpiperazine 97%
1,2-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.18 g/mol
MDL Number: MFCD08060115
CAS Number: 25057-77-6
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR