8-Bromo-6-fluoroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A243123-100MG A243123-250MG
77193-416EA 62.83 USD
77193-416 77193-418
8-Bromo-6-fluoroimidazo[1,2-a]pyridine 97%
8-Bromo-6-fluoroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
MDL Number: MFCD20923664
CAS Number: 1368664-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR