3-Fluoro-2-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

A242930-10G A242930-1G A242930-25G A242930-5G
77193-400EA 79 USD
77193-400 77193-402 77193-404 77193-406
3-Fluoro-2-methylphenylboronic acid 97%
3-Fluoro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂
MDL Number: MFCD07368679
CAS Number: 163517-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR