2-Aminoethyl acetate hydrochloride 97%

Supplier: Ambeed
Warning

A242485-1G A242485-250MG A242485-100MG
77193-384EA 151.41 USD
77193-384 77193-386 77616-000
2-Aminoethyl acetate hydrochloride 97%
2-Aminoethyl acetate hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD28139520
CAS Number: 20739-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR