Ethyl 5-iodo-1-methyl-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A241657-100MG A241657-1G A241657-250MG
77165-778EA 39.14 USD
77165-778 77165-780 77165-782
Ethyl 5-iodo-1-methyl-1H-pyrazole-4-carboxylate 97%
Ethyl 5-iodo-1-methyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉IN₂O₂
MW: 280.06 g/mol
MDL Number: MFCD21362331
CAS Number: 754219-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR