4-Methyl-5-((4-((6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
MI-463
A241508-10MG
A241508-1MG
A241508-25MG
A241508-50MG
A241508-5MG
77193-330EA
116.39
USD
77193-330
77193-332
77193-334
77193-336
77193-338
4-Methyl-5-((4-((6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile 97%
4-Methyl-5-((4-((6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-1H-indole-2-carbonitrile
Precaución: for Research Use Only
Formula:
C₂₄H₂₃F₃N₆S MW: 484.54 g/mol |
MDL Number:
MFCD28506301 CAS Number: 1628317-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...