N,3-Dimethylpyridin-2-amine 98%

Supplier: Ambeed
Warning

A241006-1G A241006-250MG
77193-320EA 254.41 USD
77193-320 77193-322
N,3-Dimethylpyridin-2-amine 98%
N,3-Dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.16 g/mol
MDL Number: MFCD16697578
CAS Number: 156267-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR