(4,6-Dichloropyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A240798-100MG A240798-1G A240798-250MG
77193-314EA 32.96 USD
77193-314 77193-316 77193-318
(4,6-Dichloropyridin-2-yl)methanol 97%
(4,6-Dichloropyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
MDL Number: MFCD26517849
CAS Number: 856163-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR