2-Phenoxy-1-phenylethanol 97%

Supplier: Ambeed
Warning

A240405-100G A240405-10G A240405-1G A240405-250MG A240405-25G A240405-5G
77165-750EA 1045.45 USD
77165-750 77165-752 77165-754 77165-756 77165-758 77165-760
2-Phenoxy-1-phenylethanol 97%
2-Phenoxy-1-phenylethanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD09880900
CAS Number: 4249-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR