4-(4-Amino-butyl)-piperazine-1-carboxylic acid tert-butyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 4-(4-aminobutyl)piperazine-1-carboxylate
A239767-100MG
A239767-250MG
A239767-1G
77193-298EA
106.09
USD
77193-298
77193-300
77746-357
4-(4-Amino-butyl)-piperazine-1-carboxylic acid tert-butyl ester 98%
4-(4-Amino-butyl)-piperazine-1-carboxylic acid tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₃H₂₇N₃O₂ MW: 257.372 g/mol |
MDL Number:
MFCD04038543 CAS Number: 745048-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...