1-Acetyl-2-imidazolidinone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Acetylimidazolidin-2-one
A239060-100G
A239060-1G
A239060-250MG
A239060-25G
A239060-1000G
A239060-5G
77165-718EA
55
USD
77165-718
77165-720
77165-722
77165-724
77165-726
77892-418
77892-419
1-Acetyl-2-imidazolidinone 97%
1-Acetyl-2-imidazolidinone
Precaución: for Research Use Only
Formula:
C₅H₈N₂O₂ |
MDL Number:
MFCD00988254 CAS Number: 5391-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...