cis-(-)-1-Phenyl-2-oxo-3-oxabicyclo[3.1.0]hexane 98%

Supplier: Ambeed
Warning

Synonyms: (1S,5R)-1-Phenyl-3-oxabicyclo[3.1.0]hexan-2-one

A239059-10G A239059-1G A239059-250MG A239059-25G A239059-5G
77165-708EA 67.98 USD
77165-708 77165-710 77165-712 77165-714 77165-716
cis-(-)-1-Phenyl-2-oxo-3-oxabicyclo[3.1.0]hexane 98%
cis-(-)-1-Phenyl-2-oxo-3-oxabicyclo[3.1.0]hexane

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MW: 174.196 g/mol
MDL Number: MFCD09833671
CAS Number: 96847-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR