17-Hydroxyheptadecanoic acid 98%

Supplier: Ambeed
Warning

A238901-1G A238901-5G A238901-250MG
77193-276EA 67.98 USD
77193-276 77193-278 77593-530
17-Hydroxyheptadecanoic acid 98%
17-Hydroxyheptadecanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₃₄O₃
MW: 286.45 g/mol
MDL Number: MFCD00506157
CAS Number: 13099-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR