5-Fluoro-2-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-methoxybenzeneboronic acid

A238244-100G A238244-1G A238244-25G A238244-5G
77193-262EA 158.62 USD
77193-262 77193-264 77193-266 77193-268
5-Fluoro-2-methoxyphenylboronic acid 98%
5-Fluoro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD01863526
CAS Number: 179897-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR