Motesanib diphosphate 98%

Supplier: Ambeed
Warning

A238109-10MG A238109-1MG A238109-50MG A238109-5MG
77193-254EA 162 USD
77193-254 77193-256 77193-258 77193-260
Motesanib diphosphate 98%
Motesanib diphosphate

Precaución: for Research Use Only

Formula: C₂₂H₂₉N₅O₉P₂
MDL Number: MFCD12407403
CAS Number: 857876-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR