Ethyl (2-Methoxybenzoyl)acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Methoxybenzoyl)acetic acid Ethyl ester, Ethyl 3-(2-Methoxyphenyl)-3-oxopropionate, Ethyl (2-methyoxybenzoyl)acetate, 3-(2-Methoxyphenyl)-3-oxopropionic acid Ethyl ester
A238009-100MG
A238009-1G
77165-694EA
30
USD
77165-694
77892-413
Ethyl (2-Methoxybenzoyl)acetate 98%
Ethyl (2-Methoxybenzoyl)acetate
Precaución: for Research Use Only
Formula:
C₁₂H₁₄O₄ |
MDL Number:
MFCD01311633 CAS Number: 41607-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...