Methyl 2-((3-fluoro-4-(methylcarbamoyl)phenyl)amino)-2-methylpropanoate 98%

Supplier: Ambeed
Warning

A237274-100MG A237274-1G A237274-250MG
77165-680EA 30.9 USD
77165-680 77165-682 77612-308
Methyl 2-((3-fluoro-4-(methylcarbamoyl)phenyl)amino)-2-methylpropanoate 98%
Methyl 2-((3-fluoro-4-(methylcarbamoyl)phenyl)amino)-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇FN₂O₃
MW: 268.28 g/mol
MDL Number: MFCD22581652
CAS Number: 1332524-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR