Benzamidine hydrochloride hydrate 98%
Supplier: Ambeed
|
Warning
|
A236159-100G
A236159-10G
A236159-25G
A236159-500G
A236159-5G
77193-220EA
52
USD
77193-220
77193-222
77193-224
77193-226
77193-228
Benzamidine hydrochloride hydrate 98%
Benzamidine hydrochloride hydrate
Precaución: for Research Use Only
Formula:
C₆H₅C(=NH)NH₂·HCl·H₂O |
MDL Number:
MFCD00066285 CAS Number: 206752-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...