(2-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A235929-100MG A235929-1G A235929-250MG
77165-666EA 37 USD
77165-666 77165-668 77165-670
(2-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid 98+%
(2-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₅BN₂O₄
MDL Number: MFCD28805507
CAS Number: 2828446-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR