4,5-Dibromo-2-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A235620-100MG A235620-1G A235620-250MG
77193-186EA 51 USD
77193-186 77193-188 77193-190
4,5-Dibromo-2-hydroxybenzaldehyde 97%
4,5-Dibromo-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Br₂O₂
MDL Number: MFCD06739554
CAS Number: 156089-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR