(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%

Supplier: Ambeed
Danger

Synonyms: (S)-2-Hydroxy-2-(trifluoromethyl)propionic Acid, (S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropanoic acid, (S)-3,3,3-Trifluoro-2-hydroxyisobutyric Acid

A234880-100MG A234880-1G A234880-250MG A234880-5G
77193-174EA 16.48 USD
77193-174 77193-176 77193-178 77193-180
(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%
(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid

Precaución: for Research Use Only

Formula: C₄H₅F₃O₃
MW: 158.08 g/mol
MDL Number: MFCD03092898
CAS Number: 24435-45-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR